L00708.22
L00708.22EA
405
EUR
InStock
L00708.22
Triphenylvinylphosphoniumbromid ≥97%
Triphenylvinylphosphoniumbromid
Purity: 97%
CAS No.: 5044-52-0
Molecular Formula: CH2=CHP(C5H5)3Br
Formula Weight: 369.25
Melting Point: 182-186°
Beilstein Reference: 4168860