L00809.03
L00809.06
L00809.14
L00809.03EA
40.5
EUR
InStock
L00809.03
L00809.06
L00809.14
3,5-Dimethoxytoluol ≥98%
3,5-Dimethoxytoluol
Purity: 98%
CAS No.: 4179-19-5
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 101°(213°F)
Refractive Index: 1.5225
Odour: Aromatic
Boiling Point: 244°
Beilstein Reference: 2043558
MDL No.: MFCD00015435