L00812.22
L00812.36
L00812.14
L00812.22EA
168
EUR
InStock
L00812.22
L00812.36
L00812.14
Biuret 97%
Biuret
Purity: 97%
CAS No.: 108-19-0
Molecular Formula: C2H5N3O2
Formula Weight: 103.08
Odour: Odourless
Melting Point: ca 189° dec.
Beilstein Reference: 1703510
Merck Reference: 14,1309
MDL No.: MFCD00007946