L01465.06
L01465.14
L01465.06EA
70.8
EUR
InStock
L01465.06
L01465.14
3-Brom-1,2,4,5-tetramethylbenzol ≥99%
3-Brom-1,2,4,5-tetramethylbenzol
Purity: 99%
CAS No.: 1646-53-3
Molecular Formula: C10H13Br
Formula Weight: 213.12
Melting Point: 58-62°
Boiling Point: 115-120°/5mm
MDL No.: MFCD00000070