L02431.06
L02431.14
L02431.06EA
45
EUR
InStock
L02431.06
L02431.14
2-Propansulfonylchlorid ≥97%
2-Propansulfonylchlorid
Purity: 97%
CAS No.: 10147-37-2
Molecular Formula: (CH3)2CHSO2Cl
Formula Weight: 142.60
Flash Point: 86°(186°F)
Refractive Index: 1.4560
Melting Point: -47°
Boiling Point: 95°/45mm
Beilstein Reference: 1747497
UN No.: 3265
MDL No.: MFCD00007453