L02513.06
L02513.14
L02513.06EA
31.7
EUR
InStock
L02513.06
L02513.14
2-(Dimethylamino)pyridin 97%
2-(Dimethylamino)pyridin
Purity: 97%
CAS No.: 5683-33-0
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 80°(176°F)
Refractive Index: 1.5590
Boiling Point: 190-192°
Beilstein Reference: 110353
MDL No.: MFCD00006261