L03071.06
L03071.14
L03071.06EA
46.4
EUR
InStock
L03071.06
L03071.14
4-Brom-o-toluylsäure ≥98%
4-Brom-o-toluylsäure
Purity: 98+%
CAS No.: 68837-59-2
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 182-184°
Beilstein Reference: 2249816