L03256.06
L03256.14
L03256.06EA
71
EUR
InStock
L03256.06
L03256.14
Ethyl-2-methylnicotinat ≥97%
Ethyl-2-methylnicotinat
Purity: 97%
CAS No.: 1721-26-2
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point: 102°(215°F)
Refractive Index: 1.5050
Odour: Recognizable
Boiling Point: 107-108°/13mm
Beilstein Reference: 118200
MDL No.: MFCD00006336