L03539.AK
L03539.AU
L03539.AKEA
24.8
EUR
InStock
L03539.AK
L03539.AU
N,N-Dimethyl-1,3-diaminopropan 99%
N,N-Dimethyl-1,3-diaminopropan
Purity: 99%
CAS No.: 109-55-7
Molecular Formula: C5H14N2
Formula Weight: 102.18
Flash Point: 32°(89°F)
Refractive Index: 1.4350
Odour: Ammonia-like
Melting Point: -70°
Boiling Point: 126-128°
Beilstein Reference: 605293
UN No.: 2734
MDL No.: MFCD00008216