L03602.06
L03602.14
L03602.06EA
77.3
EUR
InStock
L03602.06
L03602.14
Chlor(diphenyl)silan tech. 90%, Stäbe, technische Qualität
Chloro(diphenyl)silane
Purity: 90%
CAS No.: 1631-83-0
Molecular Formula: C12H11ClSi
Formula Weight: 218.76
Flash Point: 105°(221°F)
Refractive Index: 1.5790
Boiling Point: 280°
Beilstein Reference: 2937164
UN No.: 2987
MDL No.: MFCD00045144