L03992.14
L03992.22
L03992.14EA
42.3
EUR
InStock
L03992.14
L03992.22
1,4-Butansulton ≥99%
1,4-Butansulton
Purity: 99%
CAS No.: 1633-83-6
Molecular Formula: C4H8O3S
Formula Weight: 136.17
Flash Point: 153°(307°F)
Refractive Index: 1.4640
Melting Point: 12-15°
Boiling Point: 114-116°/1.5mm
Beilstein Reference: 110588
MDL No.: MFCD00006584