L04038.AE
L04038.AP
L04038.0F
L04038.AEEA
23.2
EUR
InStock
L04038.AE
L04038.AP
L04038.0F
Isobuttersäure ≥99%
Isobuttersäure
Purity: 99%
CAS No.: 79-31-2
Molecular Formula: (CH3)2CHCO2H
Formula Weight: 88.11
Flash Point: 59°(138°F)
Refractive Index: 1.3928
Odour: Pungent
Melting Point: -47°
Boiling Point: 153-154°
Beilstein Reference: 635770
Merck Reference: 14,5155
UN No.: 2529
MDL No.: MFCD00002658