Stabilization:
100 ppm Stabilisiert mit BHT/Ionol/2,6-Di-tert-butyl-4-methylphenol
L04207.09
L04207.18
L04207.09EA
50.4
EUR
InStock
L04207.09
L04207.18
2,3-Dimethyl-1,3-butadien ≥98% stabilisiert
2,3-Dimethyl-1,3-butadien
Stabilized with 100 ppm BHT
Purity: 98%
CAS No.: 513-81-5
Molecular Formula: C6H10
Formula Weight: 82.15
Storage Temperature: 2°C to 8°C
Flash Point: -1°(30°F)
Refractive Index: 1.4380
Melting Point: -76°
Boiling Point: 68-69°
Beilstein Reference: 605285
Merck Reference: 14,3238
UN No.: 3295
MDL No.: MFCD00008595