L04276.AK
L04276.AU
L04276.0F
L04276.AKEA
27.6
EUR
InStock
L04276.AK
L04276.AU
L04276.0F
Isopropylpalmitat tech. 85%, technische Qualität
Isopropylpalmitat
Purity: 85%
CAS No.: 142-91-6
Molecular Formula: CH3(CH2)14CO2CH(CH3)2
Formula Weight: 298.51
Flash Point: 183°(361°F)
Refractive Index: 1.4380
Melting Point: 11-13°
Boiling Point: 160°/2mm
Beilstein Reference: 1786567
MDL No.: MFCD00008993