L04358.AE
L04358.AP
L04358.AEEA
24.7
EUR
InStock
L04358.AE
L04358.AP
1-Chloroctan ≥99%
1-Chloroctan
Purity: 99%
CAS No.: 111-85-3
Molecular Formula: CH3(CH2)7Cl
Formula Weight: 148.68
Flash Point: 63°(145°F)
Refractive Index: 1.4298
Melting Point: -58°
Boiling Point: 182-183°
Beilstein Reference: 1697464
UN No.: 3082
MDL No.: MFCD00001022