L04387.36
L04387.36EA
100
EUR
InStock
L04387.36
Vinyltrichlorsilan ≥97%
Vinyltrichlorsilan
Purity: 97%
CAS No.: 75-94-5
Molecular Formula: C2H3Cl3Si
Formula Weight: 161.49
Storage Temperature: 2°C to 8°C
Flash Point: 10°(50°F)
Refractive Index: 1.4360
Melting Point: -95°
Boiling Point: 91-93°
Beilstein Reference: 1743440
UN No.: 1305
MDL No.: MFCD00000482