L04669.06
L04669.14
L04669.06EA
22.6
EUR
InStock
L04669.06
L04669.14
2,3-Hexandion ≥94%
2,3-Hexandion
Purity: 94%
CAS No.: 3848-24-6
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 28°(82°F)
Refractive Index: 1.4120
Odour: Characteristic
Melting Point: -30°
Boiling Point: 127-128°
Beilstein Reference: 1699896
UN No.: 1224
MDL No.: MFCD00009398