L05558.14
L05558.22
L05558.14EA
56.2
EUR
InStock
L05558.14
L05558.22
2-Phenoxyanilin ≥98%
2-Phenoxyanilin
Purity: 99%
CAS No.: 2688-84-8
Molecular Formula: C12H11NO
Formula Weight: 185.23
Flash Point: 110°(230°F)
Melting Point: 46-48°
Boiling Point: 170°/18mm
Beilstein Reference: 777766
MDL No.: MFCD00035765