L05642.03
L05642.06
L05642.03EA
67.8
EUR
InStock
L05642.03
L05642.06
3,5-Dihydroxybenzonitril ≥98%
3,5-Dihydroxybenzonitril
Purity: 98%
CAS No.: 19179-36-3
Molecular Formula: C7H5NO2
Formula Weight: 135.12
Melting Point: 185-190°
Beilstein Reference: 6322491
UN No.: 3439
MDL No.: MFCD00016453