L05715.06
L05715.14
L05715.06EA
43.3
EUR
InStock
L05715.06
L05715.14
trans-4-Decenal 96% may cont. ca 7% cis-isomer
trans-4-Decenal
Purity: 96%
CAS No.: 65405-70-1
Molecular Formula: C10H18O
Formula Weight: 154.25
Storage Temperature: 2°C to 8°C
Flash Point: 85°(185°F)
Refractive Index: 1.4450
Boiling Point: 80°/4mm
Beilstein Reference: 4230058
MDL No.: MFCD00065447