L06020.06
L06020.14
L06020.06EA
84.9
EUR
InStock
L06020.06
L06020.14
2-(4-Chlorophenyl)propionic acid 97%
2-(4-Chlorophenyl)propionic acid
Purity: 97%
CAS No.: 938-95-4
Molecular Formula: C9H9ClO2
Formula Weight: 184.62
Melting Point: 55-59°
Boiling Point: 121-126°/0.2mm
Beilstein Reference: 2328710
MDL No.: MFCD00044670