L06240.18
L06240.30
L06240.09
L06240.18EA
56.9
EUR
InStock
L06240.18
L06240.30
L06240.09
3-(2-Chlorphenyl)-5-methyl-4-isoxazolcarbonsäure ≥98%
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Purity: 98+%
CAS No.: 23598-72-3
Molecular Formula: C11H8ClNO3
Formula Weight: 237.64
Melting Point: 187-192°
Beilstein Reference: 791928