L06461.03
L06461.06
L06461.03EA
24.5
EUR
InStock
L06461.03
L06461.06
2,5-Dichlor-3-thiophensulfonylchlorid ≥97%
2,5-Dichlor-3-thiophensulfonylchlorid
Purity: 97%
CAS No.: 56946-83-9
Molecular Formula: C4HCl3O2S2
Formula Weight: 251.54
Flash Point: 110°(230°F)
Refractive Index: 1.5970
Boiling Point: 256-257°
Beilstein Reference: 1285315
UN No.: 3265
MDL No.: MFCD00051665