L06841.22
L06841.14
L06841.36
L06841.22EA
91.3
EUR
InStock
L06841.22
L06841.14
L06841.36
2,6-Diisopropylphenol ≥97%
Disoprofol
Purity: 97%
CAS No.: 2078-54-8
Molecular Formula: C12H18O
Formula Weight: 178.28
Flash Point: 113°(235°F)
Density: 0.962
Refractive Index: 1.5140
Melting Point: 17-18°
Boiling Point: 254-256°
Beilstein Reference: 1866484
Merck Reference: 14,7834
MDL No.: MFCD00008885
An anesthetic that exhibits direct activation of the GABA-A receptor