L06866.22
L06866.36
L06866.0B
L06866.XX
L06866.22EA
27.5
EUR
InStock
L06866.22
L06866.36
L06866.0B
L06866.XX
Cumolhydroperoxid 80%, technische Qualität
Cumolhydroperoxid
Purity: 80%
CAS No.: 80-15-9
Molecular Formula: C9H12O2
Formula Weight: 152.20
Storage Temperature: 2°C to 8°C
Flash Point: 56°(132°F)
Refractive Index: 1.5240
Melting Point: -30°
Boiling Point: 100-102°/8mm
Beilstein Reference: 1908117
UN No.: 3109
MDL No.: MFCD00002129