L06932.MC
L06932.ME
L06932.MCEA
30.5
EUR
InStock
L06932.MC
L06932.ME
Carbonylcyanide-3-chlorophenylhydrazone 98%
Carbonylcyanide-3-chlorophenylhydrazone
Purity: 98%
CAS No.: 555-60-2
Molecular Formula: C9H5ClN4
Formula Weight: 204.62
Storage Temperature: -30°C to -10°C
Odour: Odourless
Melting Point: ca 175° dec.
Beilstein Reference: 1842102
UN No.: UN3439
MDL No.: MFCD00001848