L07034.06
L07034.14
L07034.06EA
42.3
EUR
InStock
L07034.06
L07034.14
2-(2-Aminophenyl)benzimidazole 98%
2-(2-Aminophenyl)benzimidazole
Purity: 98%
CAS No.: 5805-39-0
Molecular Formula: C13H11N3
Formula Weight: 209.24
Melting Point: 212-216°
Beilstein Reference: 173018
MDL No.: MFCD00051496