L07130.14
L07130.22
L07130.14EA
43
EUR
InStock
L07130.14
L07130.22
(S)-(+)-Carvon 96%
(S)-(+)-Carvon
Purity: 96%
CAS No.: 2244-16-8
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 93°(199°F)
Refractive Index: 1.4980
Odour: Pleasant
Boiling Point: 228-230°
Beilstein Reference: 2042970
Merck Reference: 14,1874
Optical Rotation: +59° (neat)
MDL No.: MFCD00062997