L07360.14
L07360.06
L07360.14EA
153
EUR
InStock
L07360.14
L07360.06
2-Isopropoxyphenol ≥97%
2-Isopropoxyphenol
Purity: 97%
CAS No.: 4812-20-8
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 87°(188°F)
Refractive Index: 1.5150
Boiling Point: 100-102°/11mm
Beilstein Reference: 1937063
MDL No.: MFCD00002184