L07705.AC
L07705.AE
L07705.ACEA
48.1
EUR
InStock
L07705.AC
L07705.AE
Borwasserstoff-Dimethylsulfid-Komplex ≥94%
Borwasserstoff-Dimethylsulfid-Komplex
Purity: 94%
CAS No.: 13292-87-0
Molecular Formula: C2H9BS
Formula Weight: 75.97
Storage Temperature: 2°C to 8°C
Flash Point: -1°(30°F)
Refractive Index: 1.4570
Melting Point: -38°
Boiling Point: ca 44° dec.
Beilstein Reference: 3663489
UN No.: 2924
MDL No.: MFCD00013189