L07821.22
L07821.36
L07821.22EA
25.9
EUR
InStock
L07821.22
L07821.36
(±)-Epoxystyrol ≥98%
(±)-Epoxystyrol
Purity: 98+%
CAS No.: 96-09-3
Molecular Formula: C8H8O
Formula Weight: 120.15
Storage Temperature: 2°C to 8°C
Flash Point: 74°(165°F)
Refractive Index: 1.5350
Melting Point: -37°
Boiling Point: 193-196°
Beilstein Reference: 108582
UN No.: 2810
MDL No.: MFCD00005121