L07908.06
L07908.06EA
77.9
EUR
InStock
L07908.06
4-Chloro-3-nitrocoumarin 98%
4-Chloro-3-nitrocoumarin
Purity: 98%
CAS No.: 38464-20-9
Molecular Formula: C9H4ClNO4
Formula Weight: 225.59
Melting Point: ca 162° dec.
Beilstein Reference: 883649
MDL No.: MFCD00051670