L08015.09
L08015.18
L08015.09EA
84.7
EUR
InStock
L08015.09
L08015.18
(±)-2-Methyl-3-pentanol ≥99%
(±)-2-Methyl-3-pentanol
Purity: 99%
CAS No.: 565-67-3
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 46°(114°F)
Refractive Index: 1.4170
Boiling Point: 127-129°
Beilstein Reference: 1718952
UN No.: 2282
MDL No.: MFCD00004567