L08087.03
L08087.03EA
330
EUR
InStock
L08087.03
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol 98%
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol
Purity: 98%
CAS No.: 51-41-2
Molecular Formula: C8H11NO3
Formula Weight: 169.18
Storage Temperature: 2°C to 8°C
Melting Point: ca 220° dec.
Beilstein Reference: 4231961
Optical Rotation: -45° (c=5 in 0.5N HCl)
UN No.: UN2811
MDL No.: MFCD00025592