L08172.AE
L08172.AP
L08172.AEEA
40.5
EUR
InStock
L08172.AE
L08172.AP
Triethylorthopropionat ≥97%
Triethylorthopropionat
Purity: 98%
CAS No.: 115-80-0
Molecular Formula: C9H20O3
Formula Weight: 176.26
Flash Point: 45°(113°F)
Refractive Index: 1.4020
Odour: Slightly sweetish
Boiling Point: 158°
Beilstein Reference: 906798
UN No.: 3272
MDL No.: MFCD00009226