L08248.06
L08248.14
L08248.06EA
47.9
EUR
InStock
L08248.06
L08248.14
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) überwiegend trans ≥98%
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture)
Purity: 98+%
CAS No.: 5451-55-8
Molecular Formula: C11H20O2
Formula Weight: 184.28
Beilstein Reference: 2043187
MDL No.: MFCD00042622