L08305.03
L08305.06
L08305.14
L08305.03EA
39.6
EUR
InStock
L08305.03
L08305.06
L08305.14
(R)-(+)-1,1'-Bi-2-naphthol 99
(R)-(+)-1,1'-Bi-2-naphthol
Purity: 99%
CAS No.: 18531-94-7
Molecular Formula: C20H14O2
Formula Weight: 286.33
Odour: Characteristic
Melting Point: 206-211°
Beilstein Reference: 3616837
Merck Reference: 14,1226
Optical Rotation: +34.5 (c=1 in THF)
UN No.: 2811
MDL No.: MFCD00004068