L09432.03
L09432.06
L09432.03EA
33.4
EUR
InStock
L09432.03
L09432.06
3-Cyanphenylisocyanat ≥97%
3-Cyanphenylisocyanat
Purity: 97%
CAS No.: 16413-26-6
Molecular Formula: C8H4N2O
Formula Weight: 144.13
Storage Temperature: 2°C to 8°C
Odour: Irritating
Beilstein Reference: 3089117
UN No.: 2811
MDL No.: MFCD00013856