L09756.03
L09756.06
L09756.14
L09756.03EA
32
EUR
InStock
L09756.03
L09756.06
L09756.14
1,4-Phenylendiisothiocyanat ≥98%
1,4-Phenylendiisothiocyanat
Purity: 98%
CAS No.: 4044-65-9
Molecular Formula: C8H4N2S2
Formula Weight: 192.26
Storage Temperature: 2°C to 8°C
Odour: Irritating
Beilstein Reference: 2414340
Merck Reference: 14,1308
UN No.: 2923
MDL No.: MFCD00004811