L09811.14
L09811.22
L09811.14EA
72.5
EUR
InStock
L09811.14
L09811.22
Zinc-copper couple (∼1 - 3% Cu)
Zinc-copper couple
CAS No.: 53801-63-1
Molecular Formula: Zn/Cu
Storage Temperature: 2°C to 8°C
Melting Point: 419-420°
UN No.: UN3089
MDL No.: MFCD00084851