L09837.06
L09837.06EA
42.1
EUR
InStock
L09837.06
Brommethylmethylether tech. 90%, technical
Brommethylmethylether
Purity: 90%
CAS No.: 13057-17-5
Molecular Formula: BrCH2OCH3
Formula Weight: 124.97
Storage Temperature: 2°C to 8°C
Flash Point: 26°(78°F)
Refractive Index: 1.4520-1.4600
Odour: Pungent
Boiling Point: 86-87°
Beilstein Reference: 1730853
UN No.: 1992
MDL No.: MFCD00000171