L09875.03
L09875.06
L09875.03EA
29.5
EUR
InStock
L09875.03
L09875.06
5-Cyan-2-methylpyridin ≥99%
5-Cyan-2-methylpyridin
Purity: 99%
CAS No.: 3222-48-8
Molecular Formula: C7H6N2
Formula Weight: 118.14
Melting Point: 83-87°
Boiling Point: 216-217°
MDL No.: MFCD00038042