L09920.09
L09920.04
L09920.09EA
33.7
EUR
InStock
L09920.09
L09920.04
2-(Methylthio)nicotinsäure ≥98%
2-(Methylthio)nicotinsäure
Purity: 98+%
CAS No.: 74470-23-8
Molecular Formula: C7H7NO2S
Formula Weight: 169.20
Melting Point: 215-218°
Beilstein Reference: 4391503