L10092.09
L10092.18
L10092.09EA
61.8
EUR
InStock
L10092.09
L10092.18
3-(2-Chlor-6-fluorphenyl)-5-methyl-4-isoxazolcarbonsäure ≥99%
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Purity: 99%
CAS No.: 3919-74-2
Molecular Formula: C11H7ClFNO3
Formula Weight: 255.63
Melting Point: 201-203°
Beilstein Reference: 806523