L10150.03
L10150.06
L10150.MD
L10150.03EA
137
EUR
InStock
L10150.03
L10150.06
L10150.MD
7-(Trifluormethyl)-1,2,3,4-tetrahydrochinolin ≥97%
7-(Trifluormethyl)-1,2,3,4-tetrahydrochinolin
Purity: 97%
CAS No.: 450-62-4
Molecular Formula: C10H10F3N
Formula Weight: 201.19
Melting Point: 30-34°
Boiling Point: 135-140°/22mm
MDL No.: MFCD00079784