L10488.09
L10488.04
L10488.09EA
117
EUR
InStock
L10488.09
L10488.04
Triethyl-4-phosphonobutyrat ≥97%
Triethyl-4-phosphonobutyrat
Purity: 97%
CAS No.: 2327-69-7
Molecular Formula: C10H21O5P
Formula Weight: 252.25
Refractive Index: 1.4370
Boiling Point: 117-119°/0.4mm
MDL No.: MFCD00015168