L10532.03
L10532.06
L10532.14
L10532.03EA
21
EUR
InStock
L10532.03
L10532.06
L10532.14
Ethyl-2-piperidincarboxylat ≥98%
Ethyl-2-piperidincarboxylat
Purity: 98+%
CAS No.: 15862-72-3
Molecular Formula: C8H15NO2
Formula Weight: 157.21
Flash Point: 46°(114°F)
Refractive Index: 1.4560
Boiling Point: 216-217°
Beilstein Reference: 81694
UN No.: 1993
MDL No.: MFCD00005980