L10586.03
L10586.06
L10586.03EA
28.4
EUR
InStock
L10586.03
L10586.06
2-(Trifluormethyl)phenylisothiocyanat ≥98%
2-(Trifluormethyl)phenylisothiocyanat
Purity: 98%
CAS No.: 1743-86-8
Molecular Formula: C8H4F3NS
Formula Weight: 203.19
Flash Point: 105°(221°F)
Refractive Index: 1.5660
Odour: Pungent
Boiling Point: 216°
Beilstein Reference: 2693552
UN No.: 2922
MDL No.: MFCD00039644