L10689.06
L10689.03
L10689.06EA
124
EUR
InStock
L10689.06
L10689.03
trans-5-Decen ≥97%
trans-5-Decen
Purity: 97%
CAS No.: 7433-56-9
Molecular Formula: C10H20
Formula Weight: 140.27
Flash Point: 46°(114°F)
Refractive Index: 1.4240
Boiling Point: 171-172°
Beilstein Reference: 1719487
UN No.: 3295
MDL No.: MFCD00009508