L10777.06
L10777.14
L10777.22
AB173127.0025
L10777.06EA
40.4
EUR
InStock
L10777.06
L10777.14
L10777.22
ABCRAB173127.0025
Hexafluoraceton Trihydrat ≥98%
Hexafluoraceton Trihydrat
Purity: 98%
CAS No.: 34202-69-2
Molecular Formula: C3F6O.3H2O
Formula Weight: 220.07 (166.02anhy)
Refractive Index: 1.3190
Melting Point: 18-21°
Boiling Point: 105-106°
Beilstein Reference: 607236
UN No.: 2552
MDL No.: MFCD00149061